Learn More
Invitrogen™ FAM110B Polyclonal Antibody

Description
Immunogen sequence: KYVKSQEVIN AKQEPVKPAV LAKPPVCPAA KRALGSPTLK VFGNHAKTES GVQRENLKLE ILKNIINSSE GSSSGSGHKH SSRNWPPHRS EATDLHRHSF AESLKVYPTQ GRRSPQEGGS HVGRRLLEQS AESFLHVSHS SS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
Specifications
Specifications
| Antigen | FAM110B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAM110B |
| Gene Accession No. | Q8TC76 |
| Gene Alias | 1700012H17Rik; 5031405P22; AW049855; C8orf72; FAM110B; family with sequence similarity 110 member B; family with sequence similarity 110, member B; protein FAM110B |
| Gene Symbols | FAM110B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.