Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM110B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552543 Supplier Invitrogen™ Supplier No. PA552543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVTSVKPLKA IPCSSSAPPL PPKPKIAAIA SMKSPEADPV EPACGVSRRP SLQRSKSDLS DRYFRVDADV ERFFNYCGLD PEELENLGME NFARANSDII SLNFRSASMI SSDC Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

May be involved in tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM110B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM110B
Gene Accession No. Q5BJX5, Q8C739, Q8TC76
Gene Alias 1700012H17Rik; 5031405P22; AW049855; C8orf72; FAM110B; family with sequence similarity 110 member B; family with sequence similarity 110, member B; protein FAM110B
Gene Symbols FAM110B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM110B. Recombinant protein control fragment (Product #RP-89731).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242297, 500400, 90362
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.