Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM110C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546021
Description
Peptide sequence: DSLIIYRQKC EFVRGSGADG PRASLVKKLF QGPGKDKAPV PRTGDEGKAG Sequence homology: Human: 100%.
FAM110C may play a role in microtubule organization.
Specifications
| FAM110C | |
| Polyclonal | |
| Unconjugated | |
| FAM110C | |
| FAM110C; family with sequence similarity 110 member C; family with sequence similarity 110, member C; protein FAM110C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 642273 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q1W6H9 | |
| FAM110C | |
| Synthetic peptide directed towards the N-terminal of human FAM110C (aa 98-147). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction