Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM111A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558827 Supplier Invitrogen™ Supplier No. PA558827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSRKHSVNEK CNMKIEHYFS PVSKEQQNNC STSLMRMESR GDPRATTNTQ AQRFHSPKKN PEDQTMPQNR TIYVTLKVNH RRNQDMKL Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 47%.

With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and isconsidered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene isimportant for regulation ofcellcycle arrest and apoptosisfollowing double strand DNA breaks. Atmmutation leadsto the disorderknown as ataxia telangiectasia. The blood disorders Sickle cell anemia and β thalassemia are caused by HBB gene mutations. Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitzsyndrome are also associatedwith defectsin chromosome 11. The FAM111A gene product has been provisionally designated FAM111A pending furthercharacterization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM111A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM111A
Gene Accession No. Q96PZ2
Gene Alias 4632417K18Rik; AW413625; Fam111a; family with sequence similarity 111 member A; family with sequence similarity 111, member A; GCLEB; KCS2; Kiaa1895; Protein FAM111A; RGD1560913
Gene Symbols FAM111A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM111A. Recombinant protein control fragment (Product #RP-97448).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.