Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM111B Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297985MFV450
Description
FAM111B Polyclonal antibody specifically detects FAM111B in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
FAM111B | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
family with sequence similarity 111, member B | |
Produced in rabbits immunized with E. coli-derived Human FAM111B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
374393 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction