Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM114A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564423 Supplier Invitrogen™ Supplier No. PA564423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSDDAGDTLA TGDKAEVTEM PNSDSLPEDA EVHCDSAAVS HEPTPADPRG EGHENAAVQG AG Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 47%.

May play a role in neuronal cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM114A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM114A1
Gene Accession No. Q8IWE2
Gene Alias 1190001N04Rik; 9130005N14Rik; FAM114A1; family with sequence similarity 114 member A1; family with sequence similarity 114, member A1; nervous system overexpressed protein 20; nervous system over-expressed protein 20; Noxp20; Protein FAM114A1; Protein NOXP20; RGD1560201
Gene Symbols FAM114A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM114A1. Recombinant protein control fragment (Product #RP-103857).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.