Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM114A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114284 Supplier Invitrogen™ Supplier No. PA5114284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WVEEDQTVVS VDVAKVSEEE TKKEEKEEKS QDPQEDKKEE KKTKTIEEVY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 79%; Goat: 100%; Human: 100%; Pig: 82%; Rat: 86%; Yeast: 91%.

May play a role in neuronal cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM114A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM114A1
Gene Accession No. Q8IWE2
Gene Alias 1190001N04Rik; 9130005N14Rik; FAM114A1; family with sequence similarity 114 member A1; family with sequence similarity 114, member A1; nervous system overexpressed protein 20; nervous system over-expressed protein 20; Noxp20; Protein FAM114A1; Protein NOXP20; RGD1560201
Gene Symbols FAM114A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human FAM114A1 (aa 395-444).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92689
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.