Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM114A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 216381AP150UL

Description
May play a role in neuronal cell development.Specifications
FAM114A1 | |
Polyclonal | |
Unconjugated | |
FAM114A1 | |
FAM114A1, Noxp20, Protein FAM114A1, Protein NOXP20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
498366, 68303, 92689 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IWE2, Q9D281 | |
Fam114a1 | |
FAM114A1 Fusion Protein Ag16287 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction