Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM115A Polyclonal Antibody

Catalog No. PIPA535881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535881 Supplier Invitrogen™ Supplier No. PA535881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (94%), Bovine (98%).

TCAF1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM115A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TCAF1
Gene Accession No. Q9Y4C2
Gene Alias Fam115a; family with sequence similarity 115, member A; KIAA0738; LOC100723292; protein FAM115A; RGD1565474; Tcaf1; TRP channel-associated factor 1; TRPM8 channel associated factor 1; TRPM8 channel-associated factor 1
Gene Symbols TCAF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human FAM115A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9747
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.