Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM115C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558529 Supplier Invitrogen™ Supplier No. PA558529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WLARGQTGKV GVNTNLKDLC PLLSEHGLQC SLEPHLNSDL CVYCCKAYSD KEAKQLQEFV AE Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 61%.

The FAM115C gene (Family with Sequence Similarity 115 Member C) is a relatively under-characterized gene located on chromosome 9q34.13. This gene encodes a protein that is part of the FAM115 family, which is involved in various cellular processes, including signal transduction, cellular growth, and maintenance. While detailed functional studies on FAM115C are limited, proteins in this family are often characterized by their involvement in cellular homeostasis and regulatory mechanisms. Research on FAM115C is nascent, but it is suggested to play a role in the central nervous system based on expression patterns in brain tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM115C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCAF2
Gene Accession No. A6NFQ2
Gene Alias FAM115C; FAM139A; family with sequence similarity 115, member C; family with sequence similarity 139, member A; protein FAM115C; TCAF2; TRP channel-associated factor 2; TRPM8 channel associated factor 2; TRPM8 channel-associated factor 2
Gene Symbols TCAF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM115C. Recombinant protein control fragment (Product #RP-98828).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.