Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM116A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564469 Supplier Invitrogen™ Supplier No. PA564469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETVDLVLKLK NKLLQADREH LPVKPDTMEK LRTHIDAIIL ALPEDLQGIL L Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 96%.

Guanine nucleotide exchange factor (GEF) for RAB14. Component of an endocytic recycling pathway that is required for the control of ADAM10 transport, shedding of N-cadherin/CDH2 by ADAM9 or ADAM10 and regulation of cell-cell junctions. Required for RAB14 recruitment to recycling endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM116A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DENND6A
Gene Accession No. Q8IWF6
Gene Alias AFI1A; DENN domain containing 6A; DENN domain-containing protein 6A; DENN/MADD domain containing 6A; DENND6A; FAM116A; family with sequence similarity 116, member A; protein DENND6A; protein FAM116A
Gene Symbols DENND6A
Host Species Rabbit
Immunogen Recombinant protein fragment of Human DENND6A (Product #RP-103775)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201627
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.