Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM117B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551869 Supplier Invitrogen™ Supplier No. PA551869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECSPKQLHEI PAFYCPDKNK VNFIPKSGSA FCLVSILKPL LPTPDLTLKG SGHSLTVTTG MTTTLLQPIA VASLSTNTEQ DRVSRGTSTV MPSASLLPPP EPIEEAE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

FAM117B is encoded by ALS2CR13 which is mapped in the genomic region covering the complete candidate region for Amyotrophic lateral sclerosis 2 (ALS2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM117B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM117B
Gene Accession No. Q6P1L5
Gene Alias 2810425F24Rik; 6330416D14Rik; Als2cr13; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 13; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 13 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 13 protein homolog; FAM117B; family with sequence similarity 117 member B; family with sequence similarity 117, member B; protein FAM117B
Gene Symbols FAM117B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM117B. Recombinant protein control fragment (Product #RP-101060).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.