Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM118A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113465
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113465 Supplier Invitrogen™ Supplier No. PA5113465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM118A Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM118A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM118A
Gene Accession No. Q9NWS6
Gene Alias bK268H5.C22.4; C22orf8; FAM118A; family with sequence similarity 118 member A; family with sequence similarity 118, member A; protein FAM118A
Gene Symbols FAM118A
Host Species Rabbit
Immunogen Recombinant Human Protein FAM118A protein (51-3μAA)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55007
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.