Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM118B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559867
Description
Immunogen sequence: KHKSDLEHFM LVRRGDVDEF KKLRENMLDK GIKVISYGDD YADLPEYFKR LTCEISTRGT SAGMVREGQL NGSSAAHSE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
May play a role in Cajal bodies formation.
Specifications
FAM118B | |
Polyclonal | |
Unconjugated | |
FAM118B | |
2310022O21Rik; 2700018L24Rik; C030004A17Rik; Fam118b; family with sequence similarity 118 member B; family with sequence similarity 118, member B; Ppp4r1l; protein FAM118B; RGD1307173 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79607 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BPY3 | |
FAM118B | |
Recombinant protein corresponding to Human FAM118B. Recombinant protein control fragment (Product #RP-97557). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction