Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM118B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113314
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113314 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113314 Supplier Invitrogen™ Supplier No. PA5113314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM118B Polyclonal Antibody for Western Blot, IHC (P), ELISA

May play a role in Cajal bodies formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM118B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM118B
Gene Accession No. Q9BPY3
Gene Alias 2310022O21Rik; 2700018L24Rik; C030004A17Rik; Fam118b; family with sequence similarity 118 member B; family with sequence similarity 118, member B; Ppp4r1l; protein FAM118B; RGD1307173
Gene Symbols FAM118B
Host Species Rabbit
Immunogen Recombinant Human Protein FAM118B protein (83-351AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79607
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.