Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM118B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983323
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3323 Supplier Bioss Supplier No. BS14702R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in Cajal bodies formation.

Specifications

Antigen FAM118B
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM118B
Gene Accession No. Q9BPY3
Gene Alias 2310022O21Rik; 2700018L24Rik; C030004A17Rik; Fam118b; family with sequence similarity 118 member B; family with sequence similarity 118, member B; Ppp4r1l; Protein FAM118B; RGD1307173
Gene Symbols FAM118B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM118B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79607
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.