Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM119A Antibody (OTI2G7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272423R
Description
FAM119A Monoclonal specifically detects FAM119A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
FAM119A | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
METTL21A | |
Full length recombinant protein of human FAM119A (NP_660323) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2G7 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
EC 2.1.1.-, FAM119A, HCA557B, Hepatocellular carcinoma-associated antigen 557b, hypothetical protein LOC151194, LOC151194, member A, methyltransferase like 21A, MGC45373 | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
151194.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction