Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM119A Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
SDP

Catalog No. 502540752
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0752 30 μL
50-167-4534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0752 Supplier OriGene Supplier No. TA503703S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM119A Monoclonal antibody specifically detects FAM119A in Human samples. It is validated for Western Blot

Specifications

Antigen FAM119A
Applications Western Blot
Classification Monoclonal
Clone OTI2D6
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pdgfd
Gene Accession No. Q9GZP0
Gene Alias 2310038H17Rik; AI464204; Fam119a; family with sequence similarity 119, member A; HCA557b; heat shock protein 70kDa lysine (K) methyltransferase; hepatocellular carcinoma-associated antigen 557b; HSPA lysine methyltransferase; HSPA-KMT; LOC102553119; methyltransferase like 21A; methyltransferase like 21A; protein N-lysine methyltransferase METTL21A; methyltransferase-like protein 21A; methyltransferase-like protein 21A; protein N-lysine methyltransferase METTL21A; Mettl21A; Protein N-lysine methyltransferase METTL21A; protein N-lysine methyltransferase METTL21A-like; protein-lysine methyltransferase METTL21A
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 19-247 of human PDGFD produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80310
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.