Learn More
Invitrogen™ FAM119A Monoclonal Antibody (OTI2G7)

Description
FAM119A Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | FAM119A |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2G7 |
| Concentration | 1.00 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | METTL21A |
| Gene Accession No. | Q8WXB1 |
| Gene Alias | 2310038H17Rik; AI464204; Fam119a; family with sequence similarity 119, member A; HCA557b; heat shock protein 70kDa lysine (K) methyltransferase; hepatocellular carcinoma-associated antigen 557b; HSPA lysine methyltransferase; HSPA-KMT; LOC102553119; methyltransferase like 21A; methyltransferase like 21A; protein N-lysine methyltransferase METTL21A; methyltransferase-like protein 21A; methyltransferase-like protein 21A; protein N-lysine methyltransferase METTL21A; Mettl21A; Protein N-lysine methyltransferase METTL21A; protein N-lysine methyltransferase METTL21A-like; protein-lysine methyltransferase METTL21A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.