Learn More
FAM119A Mouse anti-Human, Clone: OTI2G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503704

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FAM119A is a protein-lysine methyltransferase that selectively trimethylates residues in heat shock protein 70 (HSP70) family members. Contributes to the in vivo trimethylation of Lys residues in HSPA1 and HSPA8. In vitro methylates Lys-561 in HSPA1, Lys-564 in HSPA2, Lys-585 in HSPA5, Lys-563 in HSPA6 and Lys-561 in HSPA8.Specifications
FAM119A | |
Monoclonal | |
Unconjugated | |
METTL21A | |
FAM119A, HCA557b, HSPA-KMT | |
Mouse | |
Affinity Chromatography | |
RUO | |
151194 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2G7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WXB1 | |
METTL21A | |
Full length human recombit protein of human FAM119A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.