Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM119A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569875 Supplier Invitrogen™ Supplier No. PA569875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

FAM119A is a protein-lysine methyltransferase that selectively trimethylates residues in heat shock protein 70 (HSP70) family members. Contributes to the in vivo trimethylation of Lys residues in HSPA1 and HSPA8. In vitro methylates Lys-561 in HSPA1, Lys-564 in HSPA2, Lys-585 in HSPA5, Lys-563 in HSPA6 and Lys-561 in HSPA8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM119A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene METTL21A
Gene Accession No. Q8WXB1
Gene Alias 2310038H17Rik; AI464204; Fam119a; family with sequence similarity 119, member A; HCA557b; heat shock protein 70kDa lysine (K) methyltransferase; hepatocellular carcinoma-associated antigen 557b; HSPA lysine methyltransferase; HSPA-KMT; LOC102553119; methyltransferase like 21A; methyltransferase like 21A; protein N-lysine methyltransferase METTL21A; methyltransferase-like protein 21A; methyltransferase-like protein 21A; protein N-lysine methyltransferase METTL21A; Mettl21A; Protein N-lysine methyltransferase METTL21A; protein N-lysine methyltransferase METTL21A-like; protein-lysine methyltransferase METTL21A
Gene Symbols METTL21A
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human METTL21A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151194
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.