Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM120A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520800
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520800 Supplier Invitrogen™ Supplier No. PA520800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-20800 can be used with blocking peptide PEP-0914.

May participate in mRNA transport in the cytoplasm. Critical component of the oxidative stress-induced survival signaling. Activates src family kinases and acts as a scaffolding protein enabling src family kinases to phosphorylate and activate PI3-kinase. Binds RNA and promotes the secretion of IGF-II. May play a pivotal role in the progression of scirrhous-type gastric cancer by supporting cancer cell survival in environments with various oxidative stresses. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM120A
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Fam120a
Gene Accession No. Q6A0A9, Q9NZB2
Gene Alias C9orf10; Constitutive coactivator of PPAR-gamma-like protein 1; DNAPTP1; DNAPTP5; Fam120a; family with sequence similarity 120, member A; family with sequence similarity 120A; Kiaa0183; Ossa; oxidative stess-associated Src activator; oxidative stress-associated Src activator; Protein FAM120A; RGD1306643
Gene Symbols Fam120a
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the center of human FAM120A.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218236, 23196, 291019
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.