Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM120A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304449100UL
Description
FAM120A Polyclonal specifically detects FAM120A in Human samples. It is validated for Western Blot.Specifications
FAM120A | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
23196 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
C9orf10MGC133257, chromosome 9 open reading frame 10, constitutive coactivator of PPAR-gamma-like protein 1, DNA polymerase-transactivated protein 1, DNA polymerase-transactivated protein 5, DNAPTP1, DNAPTP5, family with sequence similarity 120A, KIAA0183MGC111527, OSSA, Oxidative stress-associated Src activator, Protein FAM120A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human FAM120A (NP_055427.2). | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction