Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM120B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18656625UL
Description
FAM120B Polyclonal specifically detects FAM120B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FAM120B | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
CCPGKIAA0183, Constitutive coactivator of PPARG, Constitutive coactivator of PPAR-gamma, family with sequence similarity 120B, FLJ55614, KIAA1838constitutive coactivator of peroxisome proliferator-activated receptor gamma, PGCC1dJ894D12.1, PPARG constitutive coactivator 1, PPARgamma constitutive coactivator 1, Protein FAM120B | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FAM120B | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EVPMYTGSEPRQEVPMYTGPESRQEVPMYTGPESRQEVLIRTDPESRQEIMCTGHESKQEVPICTDPISKQEDSMCTHAEINQKLPVATDFEFKLEALMCTNPEIKQEDPTNV | |
25 μL | |
Lipid and Metabolism | |
84498 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction