Learn More
Invitrogen™ FAM120B Polyclonal Antibody

Description
A suggested positive control is rat thymus tissue lysate. PA5-20801 can be used with blocking peptide PEP-0915.
Specifications
Specifications
| Antigen | FAM120B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FAM120B |
| Gene Accession No. | Q6RI63, Q96EK7 |
| Gene Alias | 4932442K08Rik; AW049283; AW413338; CCPG; Constitutive coactivator of peroxisome proliferator-activated receptor gamma; constitutive coactivator of PPARG; constitutive coactivator of PPAR-gamma; dJ894D12.1; f120b; FAM120B; family with sequence similarity 120, member B; family with sequence similarity 120B; KIAA1838; mKIAA1838; PGCC1; PPARG constitutive coactivator 1; PPARgamma constitutive coactivator 1; Protein FAM120B; RGD1310304 |
| Gene Symbols | FAM120B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.