Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM120C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561568 Supplier Invitrogen™ Supplier No. PA561568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGHGKEQTGR GSKGHKKGNK QGSSDGVSKS LELHQGRSRS QVNGNSGALI KEEKSDHRLP APSQCALSRD SNECNNGNRY LPM Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.

This gene encodes a potential transmembrane protein and lies in a region where mutations and deletions have been associated with intellectual disability and autism. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM120C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM120C
Gene Accession No. Q9NX05
Gene Alias constitutive coactivator of PPAR-gamma-like protein 2; CXorf17; D930001I21Rik; FAM120C; family with sequence similarity 120, member C; family with sequence similarity 120C; ORF34; Protein FAM120C; RGD1564253; si:dkey-206f21.2; tumor antigen BJ-HCC-21
Gene Symbols FAM120C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM120C. Recombinant protein control fragment (Product #RP-100776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.