Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM120C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987517
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7517 Supplier Bioss Supplier No. BS6810R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a potential transmembrane protein and lies in a region where mutations and deletions have been associated with intellectual disability and autism. Alternative splicing results in multiple transcript variants.

Specifications

Antigen FAM120C
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM120C
Gene Accession No. Q9NX05
Gene Alias Constitutive coactivator of PPAR-gamma-like protein 2; CXorf17; D930001I21Rik; Fam120c; family with sequence similarity 120, member C; family with sequence similarity 120C; LOC100344511; ORF34; Protein FAM120C; RGD1564253; si:dkey-206f21.2; Tumor antigen BJ-HCC-21; uncharacterized LOC100344511
Gene Symbols FAM120C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM120C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54954
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.