Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM122A Monoclonal Antibody (3E9)
GREENER_CHOICE

Catalog No. PIMA524510
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524510 Supplier Invitrogen™ Supplier No. MA524510
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAQEKMELDL ELPPGTGGSP AEGGGSGGGG GLRRSNSAPL IHGLSDTSPV FQAEAPSARR NSTTFPSRHG LLLPASPVRM HSSRLHQIKQ EEGMDLINRE TVHEREVQTA MQISHSWEES FSLSDNDVEK SASPKRIDFI PVSPAPSPTR GIGKQCFSPS LQSFVSSNGL PPSPIPSPTT RFTTRRSQSP INCIRPSVLG PLKRKCEMET EYQPKRFFQG ITNMLSSDVA QLSDPGVCVS SDTLDGNSSS AGSSCNSPAK VSTTTDSPVS PAQAASPFIP LDELSSK.

Acts as an inhibitor of serine/threonine-protein phosphatase 2A (PP2A) activity. Potentiates ubiquitin-mediated proteasomal degradation of serine/threonine-protein phosphatase 2A catalytic subunit alpha (PPP2CA). Inhibits PP2A-mediated dephosphorylation of WEE1, promoting ubiquitin- mediated proteolysis of WEE1, thereby releasing G2/M checkpoint.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM122A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FAM122A
Gene Accession No. Q96E09
Gene Alias C9orf42; FAM122A; family with sequence similarity 122A; protein FAM122A
Gene Symbols FAM122A
Host Species Mouse
Immunogen FAM122A (NP_612206.3, 1 a.a. ~ 287 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116224
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.