Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM122A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563338
Description
Immunogen sequence: AQLSDPGVCV SSDTLDGNSS SAGSSCNSPA KVSTTTDSPV SPAQAASPFI PLDE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.
Acts as an inhibitor of serine/threonine-protein phosphatase 2A (PP2A) activity. Potentiates ubiquitin-mediated proteasomal degradation of serine/threonine-protein phosphatase 2A catalytic subunit alpha (PPP2CA). Inhibits PP2A-mediated dephosphorylation of WEE1, promoting ubiquitin- mediated proteolysis of WEE1, thereby releasing G2/M checkpoint.
Specifications
FAM122A | |
Polyclonal | |
Unconjugated | |
FAM122A | |
C9orf42; FAM122A; family with sequence similarity 122A; protein FAM122A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116224 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96E09 | |
FAM122A | |
Recombinant protein corresponding to Human FAM122A. Recombinant protein control fragment (Product #RP-101581). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction