Learn More
Invitrogen™ FAM122A Polyclonal Antibody

Description
Predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | FAM122A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.38 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FAM122A |
| Gene Accession No. | Q96E09 |
| Gene Alias | C9orf42; FAM122A; family with sequence similarity 122A; protein FAM122A |
| Gene Symbols | FAM122A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.