Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM122C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569857 Supplier Invitrogen™ Supplier No. PA569857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 83%; Dog: 92%; Guinea Pig: 83%; Horse: 83%; Human: 100%; Mouse: 83%; Pig: 85%; Rabbit: 83%; Rat: 83%.

FAM122C is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM122C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM122C
Gene Accession No. Q6P4D5
Gene Alias 4930432H15Rik; 4930527G05Rik; FAM122C; family with sequence similarity 122, member C; family with sequence similarity 122C; protein FAM122C; uncharacterized protein LOC685701
Gene Symbols FAM122C
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human FAM122C.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 159091
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.