Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM124A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989797
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9797 Supplier Bioss Supplier No. BS14712R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The exact function of this protein remains unknown.

Specifications

Antigen FAM124A
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene FAM124A
Gene Accession No. Q86V42
Gene Alias FAM124A; family with sequence similarity 124 member A; family with sequence similarity 124A; Protein FAM124A
Gene Symbols FAM124A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM124A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220108
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.