Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM124B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564435 Supplier Invitrogen™ Supplier No. PA564435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDETQGPLAM TVHLLANSGH GSLLQRTLDQ LLDCICPEVR LFQVSERASP VKYCEKSHSK RSRFPGMSVL LFLHESPGED RLFRVLDSLQ HSPWQCYPTQ Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.

FAM124B is identified as a novel component of CHD7 and CHD8 containing complex. It is a mainly nuclear localized protein with a widespread expressed. Fam124B is a novel protein possibly involved in the pathogenesis of CHARGE syndrome and neurodevelopmental disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM124B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM124B
Gene Accession No. Q9H5Z6
Gene Alias A830043J08Rik; Fam124b; family with sequence similarity 124 member B; family with sequence similarity 124, member B; family with sequence similarity 124B; protein FAM124B
Gene Symbols FAM124B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM124B. Recombinant protein control fragment (Product #RP-103803).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79843
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.