Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM125A Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303414100UL
Description
FAM125A Polyclonal specifically detects FAM125A in Rat samples. It is validated for Western Blot.Specifications
FAM125A | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
93343 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
CFBP, CIN85/CD2AP family binding protein, CIN85/CD2AP family-binding protein, ESCRT-I complex subunit MVB12A, family with sequence similarity 125, member A, FLJ32495, multivesicular body subunit 12A, Protein FAM125A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human FAM125A (NP_612410.1). | |
100 μL | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction