Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM126A/Hyccin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981470
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1470 Supplier Bioss Supplier No. BS11554R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.

Specifications

Antigen FAM126A/Hyccin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM126A
Gene Accession No. Q9BYI3
Gene Alias AB030242; down regulated by Ctnnb1, a; Down-regulated by CTNNB1 protein A; down-regulated by Ctnnb1, a; Drctnnb1a; Fam126a; family with sequence similarity 126 member A; family with sequence similarity 126, member A; HCC; HLD5; HYCC1; Hyccin; Protein FAM126A
Gene Symbols FAM126A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Hyccin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84668
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.