Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM126A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560152 Supplier Invitrogen™ Supplier No. PA560152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSHGLAKTA ATVFSKSFEQ VSGVTVPHNP SSAVGCGAGT DANRFSACSL QEEKLIYVSE RTELPMKHQS GQQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 77%.

Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM126A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM126A
Gene Accession No. Q9BYI3
Gene Alias AB030242; down regulated by Ctnnb1, a; down-regulated by CTNNB1 protein A; down-regulated by Ctnnb1, a; DRCTNNB1A; Fam126a; family with sequence similarity 126 member A; family with sequence similarity 126, member A; HCC; HLD5; HYCC1; Hyccin; Protein FAM126A; RGD1562906
Gene Symbols FAM126A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM126A. Recombinant protein control fragment (Product #RP-92695).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84668
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.