Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM126A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 262431AP150UL
Description
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Specifications
| FAM126A | |
| Polyclonal | |
| Unconjugated | |
| FAM126A | |
| DRCTNNB1A, FAM126A, HCC, HLD5, HYCC1, Hyccin, Protein FAM126A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84668 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BYI3 | |
| FAM126A | |
| FAM126A Fusion Protein Ag24004 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction