Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM126B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566910 Supplier Invitrogen™ Supplier No. PA566910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INSLSLIRTAS ASSSKSFDYV NGSQASTSIG VGTEGGTNLA ANNANRYSTV SLQEDRLGQA GEGKELL Highest antigen sequence identity to the following orthologs - mouse 91%, rat 34%.

Component of a complex required to localize phosphatidylinositol 4-kinase (PI4K) to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM126B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM126B
Gene Accession No. Q8IXS8
Gene Alias C130065N10Rik; D1Ertd53e; D630010C10; Fam126b; family with sequence similarity 126 member B; family with sequence similarity 126, member B; HYCC2; Protein FAM126B
Gene Symbols FAM126B
Host Species Rabbit
Immunogen Recombinant Human FAM126B. Recombinant protein control fragment (Product #RP-107562).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285172
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.