Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM127B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546682 Supplier Invitrogen™ Supplier No. PA546682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GRVQLMKALL AGPLRPAARR WRNPIPFPET FDGDTDRLPE FIVQTSSYMF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Rat: 79%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM127B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Rtl8a
Gene Accession No. Q9BWD3
Gene Alias 1110012O05Rik; CAAX box 1 homolog A; CAAX box 1A; Cxx1a; CXX1B; FAM127B; family with sequence similarity 127 member B; family with sequence similarity 127, member B; LOC679038; mammalian retrotransposon derived 8b; mammalian retrotransposon derived protein 8A; Mar8.2A/B; MAR8A; Mart8b; protein FAM127B; Retrotransposon Gag-like protein 8A; RTL8A; similar to mammalian retrotransposon derived 8b; uncharacterized protein LOC679038
Gene Symbols Rtl8a
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FAM127B (aa 3-52).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26071
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.