Learn More
Invitrogen™ FAM127C Monoclonal Antibody (OTI1A6)

Description
FAM127C Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | FAM127C |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1A6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RTL8B |
| Gene Accession No. | Q17RB0 |
| Gene Alias | CXX1c; FAM127C; family with sequence similarity 127 member C; family with sequence similarity 127, member C; mammalian retrotransposon derived protein 8B; MAR8B; protein FAM127C; Retrotransposon Gag-like protein 8B; RTL8B; SIRH4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.