Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM127C Monoclonal Antibody (OTI1A6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502175S


Description
FAM127C Monoclonal antibody specifically detects FAM127C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
FAM127C | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13501 | |
Mouse | |
Affinity chromatography | |
RUO | |
8878 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A6 | |
Unconjugated | |
Sqstm1 | |
CXX1c; FAM127C; family with sequence similarity 127 member C; family with sequence similarity 127, member C; mammalian retrotransposon derived protein 8B; MAR8B; protein FAM127C; Retrotransposon Gag-like protein 8B; RTL8B; SIRH4 | |
Full length human recombinant protein of human SQSTM1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction