Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAm127C Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
SDP

Catalog No. 502541723
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1723 30 μL
50-167-3274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1723 Supplier OriGene Supplier No. TA502197S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAm127C Monoclonal antibody specifically detects FAm127C in Canine, Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen FAm127C
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2D4
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DNAJB1
Gene Accession No. P25685
Gene Alias CXX1c; FAM127C; family with sequence similarity 127 member C; family with sequence similarity 127, member C; mammalian retrotransposon derived protein 8B; MAR8B; protein FAM127C; Retrotransposon Gag-like protein 8B; RTL8B; SIRH4
Host Species Mouse
Immunogen Full length human recombinant protein of human DNAJB1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3337, 476688
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.