Learn More
FAm127C Mouse anti-Human, Clone: OTI2D4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502197

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FAM127C (RTL8B, Retrotransposon Gag Like 8B) is a Protein Coding gene. An important paralog of this gene is RTL8A.Specifications
FAm127C | |
Monoclonal | |
Unconjugated | |
RTL8B | |
CXX1c, FAM127C, MAR8B, SIRH4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
441518 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2D4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q17RB0 | |
RTL8B | |
Full length human recombit protein of human FAM127C produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.