Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM127C Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144132
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144132 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144132 Supplier Invitrogen™ Supplier No. PA5144132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM127C Polyclonal Antibody for IHC (P), ELISA

FAM127C (RTL8B, Retrotransposon Gag Like 8B) is a Protein Coding gene. An important paralog of this gene is RTL8A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM127C
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene RTL8B
Gene Accession No. Q17RB0
Gene Alias CXX1c; FAM127C; family with sequence similarity 127 member C; family with sequence similarity 127, member C; mammalian retrotransposon derived protein 8B; MAR8B; protein FAM127C; Retrotransposon Gag-like protein 8B; RTL8B; SIRH4
Gene Symbols RTL8B
Host Species Rabbit
Immunogen Recombinant Human Retrotransposon Gag-like protein 8B protein (1-113AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441518
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.