Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM129A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115650 Supplier Invitrogen™ Supplier No. PA5115650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Niban.

Regulates phosphorylation of a number of proteins involved in translation regulation including EIF2A, EIF4EBP1 and RPS6KB1. May be involved in the endoplasmic reticulum stress response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM129A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FAM129A
Gene Accession No. Q9BZQ8, Q9ESN0
Gene Alias AI256368; AU019833; C1orf24; Cell growth-inhibiting gene 39 protein; FAM129A; family with sequence similarity 129 member A; family with sequence similarity 129, member A; GIG39; NIBAN; niban apoptosis regulator 1; niban protein; Niban1; Protein FAM129A; Protein Niban; protein Niban 1; protein Niban 1; protein Niban
Gene Symbols FAM129A
Host Species Rabbit
Immunogen A synthesized peptide derived from human FAM129A(Accession Q9BZQ8), corresponding to amino acid residues E552-S602.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116496, 63912
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.