Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM129A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555803
Description
Immunogen sequence: IHPDNCEESE VSEREAQPPC PEAHGEELGG FPEVGSPASP PASGGLTEEP LGPMEGELPG EACTLTAHEG Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 45%.
Regulates phosphorylation of a number of proteins involved in translation regulation including EIF2A, EIF4EBP1 and RPS6KB1. May be involved in the endoplasmic reticulum stress response.
Specifications
FAM129A | |
Polyclonal | |
Unconjugated | |
FAM129A | |
AI256368; AU019833; C1orf24; Cell growth-inhibiting gene 39 protein; FAM129A; family with sequence similarity 129 member A; family with sequence similarity 129, member A; GIG39; NIBAN; niban apoptosis regulator 1; niban protein; Niban1; Protein FAM129A; Protein Niban; protein Niban 1; protein Niban 1; protein Niban | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116496 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZQ8 | |
FAM129A | |
Recombinant protein corresponding to Human FAM129A. Recombinant protein control fragment (Product #RP-94229). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction