Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM129C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560306 Supplier Invitrogen™ Supplier No. PA560306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HKEEYENGGH CLGSTALTGY TLLTSQREYL RLLDALCPES LGDHTQEEPD SLLEVPVSFP LFLQHPFRRH LCFSAATR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM129C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM129C
Gene Accession No. Q86XR2
Gene Alias B-cell novel protein 1; BCNP1; BCNP1 homolog; FAM129C; family with sequence similarity 129 member C; family with sequence similarity 129, member C; NIBAN3; niban-like protein 2; Protein FAM129C; Protein Niban 3
Gene Symbols FAM129C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM129C. Recombinant protein control fragment (Product #RP-99149).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.