Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM130A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FAM130A1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FAM130A1 Polyclonal specifically detects FAM130A1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FAM130A1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
81566 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
C12ORF2, C12orf22, CSRNP-2, cysteine/serine-rich nuclear protein 2, cysteine-serine-rich nuclear protein 2, FAM130A1, family with sequence similarity 130, member A1, FLJ25576, Protein FAM130A1, TAIP12, TAIP-12chromosome 12 open reading frame 22, TGF-beta-induced apoptosis protein 12 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 324-543 of human FAM130A1 (NP_110436.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title