Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM131B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570897 Supplier Invitrogen™ Supplier No. PA570897
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM131B Polyclonal Antibody for Western Blot

FAM131B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM131B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM131B
Gene Accession No. Q86XD5
Gene Alias FAM131B; family with sequence similarity 131 member B; family with sequence similarity 131, member B; KIAA0773; Protein FAM131B
Gene Symbols FAM131B
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human FAM131B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9715
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.