Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM132A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554980 Supplier Invitrogen™ Supplier No. PA554980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAHMTWLNFV RRPDDGALRK RCGSRDKKPR DLFGPPGPPG AEVTAETLLH EFQELLKEAT ERRFSGLLD Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 65%.

Insulin-sensitizing adipocyte-secreted protein (adipokine) that regulates glucose metabolism in liver and adipose tissue. Promotes glucose uptake in adipocytes and suppresses de novo glucose production in hepatocytes via the PI3K-Akt signaling pathway. Administration lead to reduction of blood glucose. Able to attenuate inflammation in fat tissue. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM132A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1QTNF12
Gene Accession No. Q5T7M4
Gene Alias Adipolin; Adipolin cleaved form; Adipolin fC1QTNF12; Adipolin fCTRP12; Adipolin full-length form; Adipolin gC1QTNF12; Adipolin gCTRP12; Adipose-derived insulin-sensitizing factor; C1q and TNF related 12; C1q and TNF related protein 12; C1q domain containing 2; C1q/TNF-related protein 12; C1QDC2; C1QTNF12; Complement C1q tumor necrosis factor-related protein 12; CTRP12; FAM132A; family with sequence similarity 132 member A; family with sequence similarity 132, member A; protein FAM132A; zgc:158358
Gene Symbols C1QTNF12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM132A. Recombinant protein control fragment (Product #RP-90538).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.